Gas detection tube for acetonitrile
Supplier: Dräger
8103121
DRAE8103121EA 0 EUR
DRAE8103121
Gas detection tube for acetonitrile
Gas Tubes
To measure the 'spot concentrations' of acetonitrile for use with an Accuro pump.
Uses include determination and measurement of concentration peaks, personal exposure in the inhalation area, detection of leaks in pipelines or gas installations, air analysis in sewers, shafts and confined spaces.