Gas detection system, X-am® 5600
Supplier: Dräger
8321050 6811960
DRAE8321050EA 1170 EUR
DRAE8321050 DRAE6811960
Gas detection system, X-am® 5600
Gas Detection Systems
Ergonomic design and infrared sensor technology, the X-am 5600 is dedicated to the measurement of up to six gases (using dual gas sensors), employing from one to four sensors with one infrared sensor and three XXS EC sensors.
  • Practical and compact design, easy to carry
  • Dependable warnings in the event of explosive atmospheres
  • Optional external pump, makes it possible to use the detector for pre-entry measurements into confined spaces
Ordering information: The basic set (DRAE8321050) is supplied without sensors, and at least one sensor is required (More Product Information