Portable single gas detector, X-am® 5100
Supplier: Dräger
8322750
DRAE8322750EA 1170 EUR
DRAE8322750
Portable single gas detector, X-am® 5100
Gas Detection Systems
The single gas detector is designed for manufacturing petrochemical products, aseptic packing or handling of rocket fuel and for handling hydrogen fluoride (HF), hydrochloric acid (HCl), hydrogen peroxide (H₂O₂) or hydrazine.
  • Slip proof and compact design for continuous and reliable measurements
  • User friendly menus with two button operation
  • Highly resistant to impacts, vibrations and electromagnetic radiation
Operated using replaceable alkaline batteries or a T4 NiMH rechargeable battery, which can be charged in the device.
Delivery information: More Product Information