Gas-tight suits, Tychem®10000 TK, with attached boots
Supplier: DuPont
TYKGEVJDYL00/L TYKGEVJDYL00/M TYKGEVJDYL00/S TYKGEVJDYL00XL TYKGEVJDYL0XXL
DUPOTYKGEVJDYL00/LEA 4030 EUR
DUPOTYKGEVJDYL00/L DUPOTYKGEVJDYL00/M DUPOTYKGEVJDYL00/S DUPOTYKGEVJDYL00XL DUPOTYKGEVJDYL0XXL
Gas-tight suits, Tychem®10000 TK, with attached boots
Disposable Garments
This encapsulated gas-tight garment with detachable Hazmax boots (sizes 6 to 14 (39 to 48)) exhibits excellent chemical barrier properties. It is made from an extremely durable fabric that is puncture- and tear-resistant, yet also supple and lightweight (<4,6 kg per garment).
  • Wide, anti-mist visor for undistorted, panoramic visibility
  • Bat-wing design to allow the wearer to withdraw an arm to attend to breathing apparatus
  • Internal, adjustable waist belt system for support and improved fit
  • Attached double gloves, five-finger, with locking cuff mechanism
More Product Information