ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright/Inverted)
Supplier: DISCOVER ECHO
PHAS0001 RVSFEW51 CUBE6001 CUBE6002 CUBE6003 CUBE6004 CUBE6005 CUBE6006 CUBE6007 CUBE6008 CUBE6009 CUBE6010 CUBE6011 CUBE6013 CUBE6014
ECHLPHAS0001EA 170 EUR
ECHLPHAS0001 630-3325 ECHLCUBE6001 ECHLCUBE6002 ECHLCUBE6003 ECHLCUBE6004 ECHLCUBE6005 ECHLCUBE6006 ECHLCUBE6007 ECHLCUBE6008 ECHLCUBE6009 ECHLCUBE6010 ECHLCUBE6011 ECHLCUBE6013 ECHLCUBE6014
ECHO Revolve 4K Hybrid Fluorescence Microscope (Upright/Inverted)
Microscopes
This 'all-in-one' hybrid microscope can be used for upright and inverted observation methods.
  • 5 MP monochrome camera - large field of view and high sensitivity for fluorescence imaging
  • Intelligent objective nosepiece - this allows the software interface to automatically recognise the objective being used
  • Z stack software module (optional) - automated acquisition of Z stacks
  • DHR (Digital Haze Reduction) sofware module (optional) - supresses image noise and blur providing crytsal clear images
  • Illumination technique: Brightfield and epi-fluorescence (phase
More Product Information