1-Hexadecanol

Supplier: EDQM

C0990000
EDQMC0990000EA 243 EUR
EDQMC0990000
1-Hexadecanol
1-Hexadecanol
Formula: H₃C(CH₂)₁₄CH₂OH
Boiling Pt: 305…330 °C (1013 hPa)
Melting Pt: 46…52 °C
Density: 0,81 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004760
CAS Number: 36653-82-4
EINECS: 253-149-0
Merck Index: 13,02037

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR