Organic reference standards, ranitidine, Ph. Eur., RS

Supplier: EDQM

R0150000 Y0000416 Y0001787
EDQMR0150000EA 243 EUR
EDQMR0150000 EDQMY0000416 EDQMY0001787
Organic reference standards, ranitidine, Ph. Eur., RS
Standards Organic Standards

Order Now

SPECIFICATIONS

Description Ranitidine hydrochloride Ranitidine impurity J, 1,1'-N-[methylenebis(sulphanediylethylene)]bis(N'-methyl-2-nitroethene-1,1-diamine) Ranitidine for impurity A identification
CAS No. 66357-59-3 1331637-48-9 66357-59-3
Formula C₁₃H₂₃ClN₄O₃S C₁₁H₂₂N₆O₄S₂ C₁₃H₂₃ClN₄O₃S

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR