Organic reference standards, Tramadol, Ph.Eur., RS
Supplier: EDQM
Y0000155 Y0000156
EDQMY0000155EA 243 EUR
EDQMY0000155 EDQMY0000156
Organic reference standards, Tramadol, Ph.Eur., RS
Standards Organic Standards
  • In Monograph 1681
  • Solubility in water: 100 to 1000 g/l