Experimentation kits, introduction to ELISA reactions
Supplier: EDVOTEK
269
EDVO269EA 153 EUR
EDVO269
Experimentation kits, introduction to ELISA reactions
Educational Materials Biology Educational Materials Physiology and Health Learning Activities
Students learn the basic principles of the Enzyme-linked Immunosorbent Assay (ELISA) using this precise and sensitive antibody-based detection kit.
  • For 10 Lab Groups
  • Complete in 45 minutes
Also required: Distilled or deionised water, 37 °C incubator, automatic micropipettes with tips, and laboratory glassware.
Delivery information: Kit includes instructions, antigens, primary and secondary antibodies, peroxide co-substrate, hydrogen peroxide, ABTS substrate, phosphate buffered saline, tubes, plates, and transfer pipettes.