Experimentation kits, purification of the restriction enzyme Eco RI
Supplier: EDVOTEK
302
EDVO302EA 274 EUR
EDVO302
Experimentation kits, purification of the restriction enzyme Eco RI
Educational Materials Biology Educational Materials Biochemistry Learning Activities
In this experiment, students purify the restriction enzyme, Eco RI. This procedure utilised an ion exchange chromatograph step for Eco RI purification. Column fractions are assayed for the enzyme using Lambda DNA and digestion products are identified by gel electrophoresis. Fractions that contain Eco RI are identified and pooled. The total and specific activities are calculated. Recommended for advanced courses.
  • For 5 lab groups
  • Complete in 3 hours
Also required: Electrophoresis tank, power supply, UV visualisation system, water bath, microcentrifuge,More Product Information