High Performance Compact Pirani Gauge, APG200
Supplier: EDWARDS HIGH VACUUM METTLER
D1G1011100
EDWAD1G1011100EA 1020 EUR
EDWAD1G1011100
High Performance Compact Pirani Gauge, APG200
Pumps Vacuum Gauges
Edwards new APG200 series high performance compact pirani gauge is the perfect vacuum measurement solution across a spectrum of applications due to its compact size, LED light ring, integrated set points and flexibility of connections/outputs. 25% reduction in volume compared to the APG100, without any loss in performance. Maximum cable length of 100 m
  • Light ring pressure indication, compact size, long filament
  • Integrated filter
  • Wide range power supply
  • Drop in compatible
  • Set point relay
  • 360 bright LED ring
Digital gauges sit in the same footprint asMore Product Information