Oils for rotary vane pumps
Supplier: EDWARDS HIGH VACUUM METTLER
H11025011 H11025013 H11025015 H11026015 H11022013 H11022015 H11025012
EDWAH11025011EA 701 EUR
EDWAH11025011 181-0380 EDWAH11025015 EDWAH11026015 181-0379 EDWAH11022015 EDWAH11025012
Oils for rotary vane pumps
Lubricants
ULTRAgrade® 15 and 19 oils result from a new process. Most manufacturers use a solvent refining process, whereas these oils are produced by a unique hydrotreating process. This process eliminates nitrogen, sulphur and oxygen and the aromatic hydrocarbons, leaving a clear base stock essentially free of impurities.
  • When combined with additives, the ULTRAgrade® gives improved vacuum performance, thermal and oxidation stability and rust inhibition
  • The anti-oxidants included in the oil enable the pump to withstand high operating temperatures without oil
More Product Information