Assembly gloves, Tegera® 9105
Supplier: EJENDALS
9105-10 9105-11 9105-12 9105-13 9105-5 9105-6 9105-7 9105-8 9105-9
EJEN9105-10EA 0 EUR
EJEN9105-10 EJEN9105-11 EJEN9105-12 EJEN9105-13 EJEN9105-5 EJEN9105-6 EJEN9105-7 EJEN9105-8 EJEN9105-9
Assembly gloves, Tegera® 9105
Gloves
Conformable and extremely supple microthan/polyester glove that offers exceptional grip.
  • Reinforced index finger and reinforced seams
  • With Velcro and reflectors
  • Ergonomically bent fingers
  • Machine washable at 40 °C
For assembly and precision engineering work that requires the highest levels of dexterity and grip.
Certifications: EN 388 (1121X); Cat. II