Cole-Parmer® SHM-250 Digi-Mantle Heaters
Supplier: Cole-Parmer
36204-01 36204-05 36204-09 36204-13 36204-15 36204-19 36204-23
ELET36204-01EA 1350 EUR
ELET36204-01 ELET36204-05 ELET36204-09 ELET36204-13 ELET36204-15 ELET36204-19 ELET36204-23
Cole-Parmer® SHM-250 Digi-Mantle Heaters
Heaters Heating Mantles
Heating mantles with interchangeable cartridges.
  • Set speed and temperature in seconds with the intuitive display
  • Complete system requires controller and heating cartridge
  • Achieve uniform temperature up to 450 °C across round bottom flask
  • Stirring speeds from 100 to 2000 min⁻¹ with excellent stability
  • Vented casing keeps heating cartridge cool to the touch
  • Modular design allows different size heating cartridges to be used on the same Digi-mantle controller
Control temperature and stirring speed independently on the Digi-Mantle digital controller. TheMore Product Information