pH Sensor, Non-glass, Memosens CPS97E
Supplier: Endress+Hauser
CPS97E-AA8BHH2 CPS97E-AA8BHH9
ENDRCPS97E-AA8BHH2EA 778 EUR
ENDRCPS97E-AA8BHH2 ENDRCPS97E-AA8BHH9
pH Sensor, Non-glass, Memosens CPS97E
Electrodes pH Electrodes
Memosens 2.0 ISFET pH sensor for polluted media in chemical processes, paper or pigment production.
  • Memosens 2.0 offers extended storage of calibration and process data, enabling better trend identification and providing a future-proof basis for predictive maintenance and enhanced IIoT services
  • Unbreakable: PEEK sensor body withstands harsh conditions ensuring product safety
  • Open aperture prevents blocking of the sensor even in media with a high dirt load
  • Chemically stable reference gel assures long-term stable measurements
  • Low maintenance: ISFET
More Product Information