Pipette calibration software, PICASO
Supplier: EPPENDORF
3113806.000 3113807.007 3113808.003 3113809.000
EPPE3113806.000EA 45.9 EUR
EPPE3113806.000 EPPE3113807.007 EPPE3113808.003 EPPE3113809.000
Pipette calibration software, PICASO
Software Laboratory Software
PICASO (PipetteCalibrationSoftware) is a program for the automatic input and processing of measuring data from the gravimetric or photometric pipette test. With PICASO, all types of pipettes and other dispensers (e.g. bottle-top dispensers) can be checked.
  • User-oriented Windows interface, multilingual
  • On-line operating instructions and help function
  • Complete storage of a measurement including all individual values
  • Transfer of the data to other Windows programs (e.g. Excel) possible
  • Supports the archiving of complete data records
  • Analytical balances (
More Product Information