Centrifuges, benchtop, ventilated/refrigerated, 5702 / 5702 R / 5702 RH (IVD marked)
Supplier: EPPENDORF
5702000.019 5703000.012 5704000.016 5702000.060 5703000.063 5704000.067 5702000.329 5703000.322 5702000.361 5703000.365 5703000.420 5703000.462 5703000.489 5703000.381 5702000.510 5702000.558
521-0020EA 0 EUR
521-0020 521-0042 521-0593 521-0125 521-0134 521-0594 521-0992 521-0993 521-1141 521-1148 521-1718 521-1942 521-2135 521-2134 EPPE5702000.510 EPPE5702000.558
Centrifuges, benchtop, ventilated/refrigerated, 5702 / 5702 R / 5702 RH (IVD marked)
Centrifuges Benchtop Centrifuges
The 5702 range of centrifuges is ideal for medical routine, clinical and biotechnology research.
  • User friendly control panel with a small number of clearly labelled function keys and easy to read LCD display
  • Speed can be set from 100 min⁻¹ to a maximum of 4400 min⁻¹
  • Automatic, motorised lid locking
  • Slow startup and braking through the SOFT function
  • Stainless steel rotor chamber (rust-proof)
  • FastTemp function for fast precooling (5702 R/RH)
  • Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH)
More Product Information