Ductless filtering fume hoods, Captair® Smart and Midcap 321
Supplier: ERLAB CAPTAIRE
AA602A00011A21 AA602A00011A31 AA602A00011A41 AA606A00011A31
ERLAAA602A00011A21EA 2830 EUR
ERLAAA602A00011A21 ERLAAA602A00011A31 ERLAAA602A00011A41 ERLAAA606A00011A31
Ductless filtering fume hoods, Captair® Smart and Midcap 321
Fume Hoods and Enclosures Ductless Fume Hoods
The Captair® Smart ductless filtering hoods are designed with a simple and innovative way of communication. Smart Technology, simple and intuitive communication by light indicates the status of the hood. A simple light pattern will indicate if the airflow is compromised, if there is a fan failure or in case of a filter breakthrough.
Please complete the quote request to enable us to assist you in finding the right unit for your application.
https://ie.cmd2.vwr.com/bin/public/fc/main?f=Y5&pimid=16984845
  • The Erlab's unique Flex Filtration can manage most laboratory applications
  • New
More Product Information