Anti-Tissue Factor Pathway Inhibitor Goat Polyclonal Antibody

Supplier: Everest Biotech
EB05048
EVBIEB05048EA 339 EUR
EVBIEB05048
Anti-Tissue Factor Pathway Inhibitor Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Tissue Factor Pathway Inhibitor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR