Anti-LAX1 Goat Polyclonal Antibody

Supplier: Everest Biotech
EB06080
EVBIEB06080EA 202 EUR
EVBIEB06080
Anti-LAX1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LAX1 (lymphocyte transmembrane adaptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR