Anti-MPP5 Goat Polyclonal Antibody

Supplier: Everest Biotech
EB06223
EVBIEB06223EA 339 EUR
EVBIEB06223
Anti-MPP5 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MPP5 (membrane protein, palmitoylated 5 (MAGUK p55 subfamily member 5))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR