Anti-VAV3 Goat Polyclonal Antibody

Supplier: Everest Biotech
EB06467
EVBIEB06467EA 339 EUR
EVBIEB06467
Anti-VAV3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: VAV3 (vav 3 guanine nucleotide exchange factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR