Anti-LXR alpha / LXR beta Goat Polyclonal Antibody

Supplier: Everest Biotech
EB06522
EVBIEB06522EA 339 EUR
EVBIEB06522
Anti-LXR alpha / LXR beta Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LXR alpha / LXR beta
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR