Anti-mKIAA0319 Goat Polyclonal Antibody

Supplier: Everest Biotech
EB07476
EVBIEB07476EA 339 EUR
EVBIEB07476
Anti-mKIAA0319 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: mKIAA0319
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR