Anti-SLC7A11 Goat Polyclonal Antibody

Supplier: Everest Biotech
EB07480
EVBIEB07480EA 339 EUR
EVBIEB07480
Anti-SLC7A11 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR