Anti-CYTH4 Goat Polyclonal Antibody

Supplier: Everest Biotech
EB08214
EVBIEB08214EA 202 EUR
EVBIEB08214
Anti-CYTH4 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CYTH4 (cytohesin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol CYTH4
Antigen name cytohesin 4
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Goat
Isotype IgG
ELISA Yes
Epitope internal
Cross adsorption none
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR