Anti-USP9X Goat Polyclonal Antibody

Supplier: Everest Biotech
EB08669
EVBIEB08669EA 339 EUR
EVBIEB08669
Anti-USP9X Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: USP9X (ubiquitin specific peptidase 9, X-linked)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR