Anti-CDON Goat polyclonal antibody unconjugated

Supplier: Everest Biotech
EB09472
EVBIEB09472EA 202 EUR
EVBIEB09472
Anti-CDON Goat polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: CDON (cell adhesion associated, oncogene regulated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR