Anti-KCNN2 Goat Polyclonal Antibody

Supplier: Everest Biotech
EB09678
EVBIEB09678EA 339 EUR
EVBIEB09678
Anti-KCNN2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KCNN2 (potassium intermediate/small conductance calcium-activated channel, subfamily N, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR