Anti-LSP1 Goat Polyclonal Antibody

Supplier: Everest Biotech
EB09870
EVBIEB09870EA 339 EUR
EVBIEB09870
Anti-LSP1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LSP1 (lymphocyte-specific protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR