Anti-NLRX1 / NOD9 Goat Polyclonal Antibody

Supplier: Everest Biotech
EB09892
EVBIEB09892EA 339 EUR
EVBIEB09892
Anti-NLRX1 / NOD9 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NLRX1 / NOD9
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR