Anti-DCUN1D1 Goat polyclonal antibody unconjugated

Supplier: Everest Biotech
EB09917
EVBIEB09917EA 339 EUR
EVBIEB09917
Anti-DCUN1D1 Goat polyclonal antibody unconjugated
Antibodies
Type: Primary
Antigen: DCUN1D1 (DCN1, defective in cullin neddylation 1, domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR