Anti-ULK3 Goat Polyclonal Antibody

Supplier: Everest Biotech
EB11184
EVBIEB11184EA 339 EUR
EVBIEB11184
Anti-ULK3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ULK3 (unc-51 like kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR