Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 3 µm

Supplier: Avantor
EXL-1111-1002UEA 1100 EUR
EXL-1111-1002U EXL-1111-1003U EXL-1111-1046U EXL-1111-1203U EXL-1111-1246U EXL-1111-1502U EXL-1111-1503U EXL-1111-1546U EXL-1111-2046U EXL-1111-0202U EXL-1111-0203U EXL-1111-0246U EXL-1111-2502U EXL-1111-2503U EXL-1111-2546U EXL-1111-0302U EXL-1111-0503U EXL-1111-0546U EXL-1111-7502U EXL-1111-7503U EXL-1111-7546U EXL-1111-0502U EXL11111502UMV EXL1111503UMVK
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 3 µm
Chromatography Columns
Avantor® ACE® Excel® SuperC18 columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.

  • Ultra-inert for maximum performance and reproducibility
  • Ultra low bleed for improved LC/MS compatibility
  • Wide range of particle sizes (1,7 to 10 µm) and complementary phases

Maximum recommended pH: 1,5 to 11,5

Max. pressure: 1000 bar
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR