Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 3 µm
Supplier: Avantor
EXL-1112-1002UEA 1090 EUR
EXL-1112-1002U EXL-1112-1003U EXL-1112-1046U EXL-1112-1202U EXL-1112-1502U EXL-1112-1503U EXL-1112-1546U EXL-1112-0246U EXL-1112-2502U EXL-1112-2503U EXL-1112-2546U EXL-1112-0302U EXL-1112-3546U EXL-1112-0502U EXL-1112-0503U EXL-1112-0546U EXL-1112-7502U EXL-1112-7503U EXL-1112-7546U
Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 3 µm
Chromatography Columns
Avantor® ACE® Excel® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1,7 to 5 µm) and complementary phases
  • Low bleed for UV and LC/MS
More Product Information