Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
EXL-122-1046UEA 948 EUR
EXL-122-1046U EXL-122-1246U EXL-122-1546U EXL-122-0202U EXL-122-0246U EXL-122-2502U EXL-122-2546U EXL-122-0502U EXL-122-0503U
Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 5 µm
Chromatography Columns
These ultra-inert Avantor® ACE® Excel® C8 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 5 µm) and complementary phases
  • Can be used as general purpose starting point column
  • Method validation kits contain three different batches of the same phase -
More Product Information