Aqueous mounting medium, Fluoroshield™ with DAPI, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

F6057-20ML
F6057-20MLEA 150 EUR
F6057-20ML
Aqueous mounting medium, Fluoroshield™ with DAPI, Sigma-Aldrich®
Stains Microscopy and staining reagents

Fluoroshield™ with DAPI is an aqueous mounting medium for preserving fluorescence of tissue and cell smears.


  • Storage temperature: 2 - 8 °C


This mounting medium is fortified with DAPI which is a counterstain for DNA. For use in in situ hybridization techniques or other methods where fluorescence of DNA staining is required. DAPI excites at 360 nm and emits at 460 nm, producing a blue fluorescence. RNA is also stained with DAPI.


Applications:
- Counterstain in immunocytochemistry to label nuclei of neural progenitor cells
- Histology mounting medium for cardiac tissues sections for immunofluorescence studies
- Mounting solution for immunostained heart sections for nuclei detection

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR