Planetary mill, PULVERISETTE 5 classic line
Supplier: FRITSCH
05.6010.00 05.5010.00
FRIT05.6010.00EA 16900 EUR
FRIT05.6010.00 FRIT05.5010.00
Planetary mill, PULVERISETTE 5 classic line
Mills
These planetary ball mills are ideal for comminution of solid matter in suspension down to colloidal fineness, crushing of dry laboratory samples, and mixing and homogenizing of emulsions and pastes. During operation, grinding bowls with material and balls rotate around their own axis on a counter-rotating supporting disc. Mills are equipped with two- or four-bowl fasteners and a 'safe-lock' clamping system.
  • Suitable for hard, medium-hard, soft, brittle, tough or moist material samples
  • Toothed belt drive for bowls provides constant transmission ratio
More Product Information