Variable-speed rotor mill, PULVERISETTE 14
Supplier: FRITSCH
14.5020.00
FRIT14.5020.00EA 8580 EUR
FRIT14.5020.00
Variable-speed rotor mill, PULVERISETTE 14
Mills
The FRITSCH variable speed rotor mill PULVERISETTE 14 classic line is the ideal mill for fast, effective comminution of soft to medium-hard, brittle and fibrous materials as well as temperature-sensitive samples.
  • Rapid sample throughput due to high-speed grinding and simple cleaning
  • Max. feed size 10 mm, max. throughput 5 L/h
  • Final fineness d50 <40 µm, sieve inserts 0,08 to 6 mm
  • Simple changing of all grinding parts without tools
  • Variable speed settings between 6000 and 20000 min⁻¹
  • Automatic speed regulation for constant milling
Ordering information:More Product Information