Anti-Cre Recombinase Mouse monoclonal antibody unconjugated

Supplier: Genscript
A00888-40
GENSA00888-40EA 192 EUR
GENSA00888-40
Anti-Cre Recombinase Mouse monoclonal antibody unconjugated
Antibodies
Type: Tag
Antigen: Cre Recombinase
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity:
Order Now

SPECIFICATIONS

Environmentally Preferable
Antibody type Tag
Clonality Monoclonal
Conjugation Unconjugated
Reactivity
Host Mouse
Isotype IgG2a
Western blot Yes
ELISA Yes
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR