Anti-TUBA Mouse Monoclonal Antibody

Supplier: Genscript
A01410-100
GENSA01410-100EA 419 EUR
GENSA01410-100
Anti-TUBA Mouse Monoclonal Antibody
Antibodies
Type: Tag
Antigen: TUBA (tubulin, alpha)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR