Anti-V5 Tag Mouse Monoclonal Antibody (Biotin)

Supplier: Genscript
A01735-100
GENSA01735-100EA 410 EUR
GENSA01735-100
Anti-V5 Tag Mouse Monoclonal Antibody (Biotin)
Antibodies
Type: Tag
Antigen: V5 Tag (V5 tag)
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR