Sand, Crystalline Ø 0.1 - 0.5 mm, washed, calcinated

Supplier: GERBER FUNKE

Synonyms: Silicium dioxide, Silicium (IV) oxide, Silicon dioxide

5460 5461 5462 5464
GERB5460EA 14 EUR
GERB5460 GERB5461 GERB5462 GERB5464
Sand, Crystalline Ø 0.1 - 0.5 mm, washed, calcinated
Sand

Crystalline, calcined quality.

Formula: SiO₂
MW: 60.08 g/mol
Melting Pt: 1700
Density: 2.6
MDL Number: MFCD02100519
CAS Number: 7631-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR