Silicone encased USB computer mouse for hygienic areas, GCQ® PrimeMouse
Supplier: GETT GERATETECHNIK
KH29213 KH29214
GETTKH29213EA 64.8 EUR
GETTKH29213 GETTKH29214
Silicone encased USB computer mouse for hygienic areas, GCQ® PrimeMouse
Computer Mice and Mouse Pads
The PrimeMouse offers ergonomic functionality with two mouse buttons and 3 scroll buttons. Due to its symmetrical housing shape, it is perfectly suitable for both right-handed and left-handed users. The PrimeMouse also has a completely waterproof and dustproof silicone housing.
  • Ergonomical functionality with two mouse buttons and three scroll buttons
  • Symmetrical shaped silicone housing is suited for right and left hand operation
  • IP 68 Silicone mouse with fully waterproof and dustproof housing
  • Optical sensor with a resolution of 1000 dpi
  • With plastic base
More Product Information