Aqueous mounting medium, glycerol gelatin, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

GG1-10X15ML GG1-15ML
GG1-10X15MLEA 206 EUR
GG1-10X15ML GG1-15ML
Aqueous mounting medium, glycerol gelatin, Sigma-Aldrich®
Stains Microscopy and staining reagents

Glycerol alters the hygroscopic property of gelatin and its permeability to water vapour. Glycerol gelatin or glycerine jelly medium is an aqueous slide mounting media, used for mounting samples in microscopy and histology.


  • Storage temperature: 2 - 8 °C


Applications: Glycerol Gelatin has been used for mounting endometrial tissue and pancreatic sections for microscopic examination. It has also been used for the immobilization and mounting of helminth eggs for on chip image analysis.


Prepared with gelatin, glycerol and phenol. Very similar in composition to Kaiser′s glycerol jelly.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR