Cryoboxes, PP, D270
Supplier: GLW
D270
GLWAD270EA 8.6 EUR
GLWAD270
Cryoboxes, PP, D270
Boxes Cryoboxes
Cryoboxes are available in various heights. It is possible to customize the box according to individual storage needs.
  • Made from polypropylene
  • Autoclavable up to +121 °C and use in freezer down to –90 °C
  • The included clear lid offers a view of the contents without opening the cryobox
  • Without drainage holes in base for catching drips or store fluids (leakproof only at bottom up to 40 mm)
  • Height extension secured with exclusive clicksystem (not glued)
The box with drainage holes is ideal for rock samples and bulky items, for example. The GLW box isMore Product Information