IIRR410 LG 102030415W1WW FRIDGE, white exterior * refrigerant R600a * RIGHT hinged SOLID door * castors 125mm * e-sensor * 5 white wire shelves 430mm * 1 white wire shelf 300mm 1 * 1 items
Supplier: GRAM
IIRR410 LG 102030415W1WW FRIDGE, white exterior * refrigerant R600a * RIGHT hinged SOLID door * castors 125mm * e-sensor * 5 white wire shelves 430mm * 1 white wire shelf 300mm 1 * 1 items
Learn more About VWR
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.