Half area microplates, 96-well
Supplier: Greiner Bio-One
675075_40 675101_40 675074_40 675076_40 675077_40 675096_40 675161_40
392-0287EA 220 EUR
392-0287 738-0145 GREI675074_40 392-0288 GREI675077_40 392-0290 GREI675161_40
Half area microplates, 96-well
Microplates HTS Microplates
PS, without lid. 96-well half area microplates offer an alternative to 384-well or 1536-well microplates when low sample volume is an important criterion but automation is not necessarily appropriate. They can be pipetted manually while at the same time allow a reduction in sample volume of up to 50%.
Microplates with clear bottoms feature micro-thin µClear® bottoms for exceptional optical properties and standardised pathlength, eliminating the need for pathlength determinations. White plates are ideal for luminescence assays, while black plates are suitable for fluorescence assays. BothMore Product Information