Urine Cups with Integrated Transfer Device
Supplier: Greiner Bio-One
724321 724322
GREI724321EA 166 EUR
GREI724321 GREI724322
Urine Cups with Integrated Transfer Device
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Urine Sample Collection/Preparation
The Urine Cup with integrated transfer device allows you to collect and transport urine samples safely to the necessary destination. Thanks to the integrated transfer device, sample transfer from the cup to an evacuated tube is simple, fast and hygienic.
  • Recessed transfer device to reduce risk of needle stick injuries
  • Available bulk packed or in cases of small packs to suit different distribution needs within healthcare facilities
The optimised threading of the urine cup makes it easy to open and close. In addition, a stabilising flange makes the cupMore Product Information