2-Hydroxy-4-(trifluoromethyl)pyridine ≥98%

Supplier: Thermo Fisher Scientific
Warning

H25756.06
H25756.06EA 0 EUR
H25756.06
2-Hydroxy-4-(trifluoromethyl)pyridine ≥98%
2-Hydroxy-4-(trifluoromethyl)pyridine

Purity: 98% CAS No.: 50650-59-4 Molecular Formula: C6H4F3NO Formula Weight: 163.10 Melting Point: 158-160° MDL No.: MFCD00153195

Formula: C₆H₄F₃NO
Melting Pt: 158…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00153195
CAS Number: 50650-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR